Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Serum, Type AB

Catalog No. ICN2938249 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2938249 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2938249 Supplier MP Biomedicals™ Supplier No. 092938249
Add to Cart
Add to Cart

Golden Yellow Liquid. Proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Color Golden Yellow
Content And Storage -20°C
Species Human
Form Liquid
Product Type Human Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.1 μm filtration
For Use With (Application) Grow many Human cell lines at a faster rate
Heat Inactivated Yes
Hemoglobin Concentration ≤26mg/dL
Mycoplasma Detected No
Osmolality 263 to 323mOsm/kg
pH 7 to 8
Protein Content 5.5 to 7.5 g/dL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.