Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hyaluronidase, From Streptomyces hyalurolyticus

Catalog No. p-96381 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
200 U
Inhibitors:
Mn2+, Hg2+, K+ Ferricyanide
Molecular Weight (g/mol):
55
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Inhibitors Molecular Weight (g/mol)
ICN15127080 100 U Mn2+, Hg2+, K+ Ferricyanide 55
ICN15127082 200 U Mn2+, Hg2+, K+ Ferricyanide 55
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15127080 Supplier MP Biomedicals™ Supplier No. 0215127080
Only null left
Add to Cart
Add to Cart

Enzyme activity is determined by a change in optical density in a hyaluronic acid solution.

  • From Streptomyces hyalurolyticus
  • Lyophilized, salt-free
  • Activity: ∽2000 units/mg
  • Protease and lysozyme free
  • Each vial contains 100U

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈2000 U/mg
Color White
pH 6
For Use With (Application) Increase the absorption and dispersion of injected drugs or to disrupt the structure of certain organs
Content And Storage 4°C
Source Streptomyces hyalurolyticus
Quantity 100 U
Product Type Hyaluronidase
Molecular Weight (g/mol) 55
Inhibitors Mn2+, Hg2+, K+ Ferricyanide
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.