Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hyaluronidase, Source: Ovine Testes

Catalog No. ICN15127201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15127201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15127201 Supplier MP Biomedicals™ Supplier No. 0215127201

missing translation for 'validMainframeMsg'

Ovine Testes

Hyaluronidase, [37326-33-3]

  • Hyaluronate Glycanohydrolase
  • Lyophilized, Salt-free
  • Activity: 500 I.U./mg material

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 4.5 to 6
For Use With (Application) Drug dispersion and delivery
Content And Storage -20°C
Source Ovine testes
Quantity 1 g
Product Type Hyaluronidase
Molecular Weight (g/mol) ≈55
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.