Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hyaluronidase, Streptomyces hyalurolyticus

Catalog No. ICN15127084 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
400 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15127084 400 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15127084 Supplier MP Biomedicals™ Supplier No. 0215127084
Add to Cart
Add to Cart

Source: Streptomyces hyalurolyticus

Hyaluronidase is the enzyme that degrades hyaluronic acid in mucopolysaccharides to low molecular saccharides. Hyaluronidase purified from Streptomyeshyalurolyticus is distinct from the known hyaluronidase in its substrate specificity and mechanism of action. This enzyme is quite specific for hyaluronic acid. Various types of chondroitin sulfate (A,B,C,D,,E), chondroitin, heparin, keratan sulfate and chitin are not attacked by this enzyme. This enzyme splits the N-acetylglucosaminic linkages in hyaluronic acid eliminatively , yielding ▿-4,5-unsaturated tetrasaccharides and hexasaccharides as the end-product.

  • Formula Weight: 55kDa
  • CAS #: 37326-33-3
  • Synonyms: EC 4.2.2.1; Hyaluronate lyase; Mucinase
  • Physical Appearance : White lyophilized powder
  • Typical : ã 2,000 TRU/mg protein
  • Contaminants : Protease activity: less than 0.05 u/ampule. One unit is the amount of protease activity which produces 100 ++g of tyrosine from milk casein at 37°C for 60 min. (pH 6.0)
  • Optimum pH: 6.0
  • Recommended reaction temperature : 60°C
  • Molecular Weight : 91,000 (gel filtration)
  • Inhibitors : Mn2+, Hg2+, Potassium ferricyanide
  • Unit Definition : One TRU (Turbidity Reducing Unit) is that amount of enzyme which causes 50% of decrease in absorbance at 660 nm in 30 minutes at 60°C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈2000 U/mg
Color White
pH 6
For Use With (Application) Used to increase the absorption and dispersion of injected drugs or to disrupt the structure of certain organs
Content And Storage 4°C
Source Streptomyces hyalurolyticus
Quantity 400 U
Product Type Hyaluronidase
Molecular Weight (g/mol) 91000
Inhibitors Mn2+, Hg2+, Potassium Ferricyanide
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.