Learn More
Description
- Format: Tan powder
- Specific Activity: Standard quality of contains at least 50 Coli units per mg (total binding)
Absorption of vitamin B12
Specifications
Specifications
| For Use With (Application) | Vitamin B12 Absorption |
| Name | Intrinsic Factor from Porcine Pyloric Mucous Lining |
| Quantity | 100 mg |
| Storage Requirements | 0°C |
| Form | Powder |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.