Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lactase from Aspergillus oryzae

Catalog No. p-4605366 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10078001 1 g
ICN10078025 25 g
ICN10078005 5 g
ICN10078080 100 g
ICN10078083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10078001 Supplier MP Biomedicals™ Supplier No. 0210078001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaves lactose into its monosaccharide components, glucose and galactose

  • Hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and α-D-Glucose
  • Catalyses transglycosylation of glucose into allolactose, the inducer of β-galactosidase

  • EC Number: 232-864-1
  • MDL Number: MFCD00130623
  • Format: Off white powder
  • Specific Activity: >5000U/g
  • Stability: Stable in pH range of 4.0 to 8.0
  • Stable to heat
  • Soluble in water, insoluble in ethanol, acetone and isopropyl alcohol
  • Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate

Used to screen for blue white colonies in MCS of various plasmid vectors in Escherichia coli or other bacteria and to break down lactose in milk to make it suitable for people with lactose intolerance.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >5000 U/g
Color Off-White
pH 4 to 8
For Use With (Application) Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk
Content And Storage 4°C
Source Aspergillus oryzae
Quantity 1 g
Product Type Lactase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.