Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Linbro™ PCR Rainbow Microcentrifuge Tubes

Catalog No. ICN15950190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Color:
Assorted
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
ICN15950190 Assorted
ICN15950090 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15950190 Supplier MP Biomedicals™ Supplier No. 0215950190

missing translation for 'validMainframeMsg'

Use these V-shaped Plastic Snap Top Tubes for PCR applications

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 0.25 mL
Color Assorted
Material Polypropylene
For Use With (Application) PCR applications
Includes Frosted writing area
Packaging 500 Case
Bottom Shape Conical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.