Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lipase from Porcine Pancreas

Catalog No. ICN10081701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10081701 1 g
ICN10081710 10 g
ICN10081705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10081701 Supplier MP Biomedicals™ Supplier No. 0210081701
Add to Cart
Add to Cart

Catalyzes hydrolysis of esters in aqueous solutions and synthesis of esters in non-aqueous solutions

Lipases are a subclass of esterases. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

  • Synonyms: Steapsin, Glycerol ester hydrolase, Tween hydrolase, Tributyrase, PPL, Triacylglycerol acylhydrolase, Triacylglycerol lipase
  • CAS Number: 9001-62-1
  • EC Number: 232-619-9
  • MDL Number: MFCD00131509
  • Format: Tan powder
  • Specific Activity: >12,000U/g solid
  • Extinction Coefficient: E1%280=13.3
  • Isoelectric point (pI): Lipase A=4.9 and Lipase B=5.0
  • Partly soluble in water and insoluble in alcohol

Used industrially for resolution of chiral compounds and transesterification production of biodiesel.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Chiral Compounds Resolution, Transesterification Production of Biodiesel
Name Lipase
Quantity 1 g
Source Porcine Pancreas
Storage Requirements 0°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.