Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysozyme from Chicken Egg White

Catalog No. ICN10083410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10083410 10 g
ICN10083405 5 g
ICN10083401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10083410 Supplier MP Biomedicals™ Supplier No. 0210083410

2X crystallized, salt-free and soluble

Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.

  • Lyophilized
  • Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
  • CAS: 9001-63-2

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color White
pH 11
For Use With (Application) Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen
Content And Storage 0°C
Source Chicken egg white
Quantity 10 g
Product Type Lysozyme
Molecular Weight (g/mol) 14388
Inhibitors Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes
Form Crystallized, Lyophilized
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.