Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Malate Dehydrogenase, From Porcine Heart (Mitochondrial)

Catalog No. ICN16004910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16004910 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN16004910 Supplier MP Biomedicals™ Supplier No. 0216004910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for enzymatic determination of L-malate and gluamate oxaloacetate transaminase (GOT)

Malate Dehydrogenase, [9001-64-3]

  • CAS Number: 9001-64-3
  • EC Number: 232-622-5
  • Specific Activity: ∽1200u/mg protein
  • Format: Powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈1200 U/mg
Color White
For Use With (Application) Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase
Content And Storage 0°C
Source Porcine heart
Quantity 10 kU
Product Type Malate Dehydrogenase
Form Lyophilized Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.