Learn More
MP Biomedicals™ Microplate Sealing Tape
Catalog No. ICN7742000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Roll
Catalog No. Quantity
ICN7742000 1 Roll
1 options
Catalog No. ICN7742000 Supplier MP Biomedicals™ Supplier No. 097742000
Add to Cart
Add to Cart
Suitable for all microplates for both short- and long-term storage, providing a tight seal to minimize evaporation and condensation.
TRUSTED_SUSTAINABILITY
Specifications
Material Acetate
No. per Case Each
Quantity 1 Roll
For Use With (Application) Suitable for all microplates for both short and long term storage, providing a tight seal to minimize evaporation and condensation
Product Type Microplate Sealing Tape
WARNING: Cancer - www.P65Warnings.ca.gov