Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Molecular Genetics Agar

Catalog No. ICN19402283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19402283 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19402283 Supplier MP Biomedicals™ Supplier No. 0219402283

missing translation for 'validMainframeMsg'

Suitable as a component in culture media for molecular genetics

Powder. CAS Reg. No.: 9002-18-0.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Agar
For Use With (Application) Suitable as a component in culture media for molecular Genetics
Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.