Learn More
Description
- Formula Weight: 737.87
- CAS # 8005-03-6
- Synonym: Acid Black 2; C.I. 50420
- Physical Description: Black crystalline powder
- Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
Specifications
Specifications
| Quantity | 25 g |
| Format | Crystal |
| For Use With (Application) | Viable cell staining agent especially in stem cell biology |
| Storage Requirements | 15 to 30°C |
| Product Type | Nigrosin |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.