Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pancreatin 8X, USP Grade

Catalog No. ICN19397625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19397625 25 g
ICN19397690 500 g
ICN19397680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19397625 Supplier MP Biomedicals™ Supplier No. 0219397625
Add to Cart
Add to Cart

From porcine pancreas

Composed of numerous enzymes including amylase, trypsin, lipase, ribonuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in five minutes in water at 40°C, and not less than than 25 times its weight of casein into proteoses. It hydrolyzes fats to glycerol and fatty acid, protein to proteoses and derived compounds, and starch into dextrins and sugar. Lipase Activity: ≥16U/mg. Amylase Activity: ≥200U/mg. Protease Activity: ≥200U/mg.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 8X
Color Cream
For Use With (Application) Promote esophageal tissue loosening in patients with esophageal tumors
Content And Storage 0°C
Source Porcine pancreas
Quantity 25 g
Product Type Pancreatin
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.