Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Papain from Papaya Latex

Catalog No. ICN10092125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
500 mg
1 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10092125 25 mg
ICN10092180 100 mg
ICN10092150 50 mg
ICN10092190 500 mg
ICN10092191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10092125 Supplier MP Biomedicals™ Supplier No. 0210092125
Add to Cart
Add to Cart

Sulfhydryl protease from Carica papaya latex

Papain cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.

  • CAS Number: 9001-73-4
  • EC Number: 232-627-2
  • MDL Number: MFCD00131791
  • Format: Milky white solution
  • Specific Activity: ≤10 units/mg Protein
  • Extinction coefficient: 25.0
  • Isoelectric point (pI): pH 9.6
  • Optimum pH: 6.0 to 7.0

Used to produce Fab fragments of antibodies, cell dissociation, morphological isolation of intact cortical neurons from postnatal rats. Limited papaindigestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify red cell surface to enhance or destroy reactivity of many red cell antigens. Useful for platelet serology studies and has also been used in enzymatic synthesis of amino acids, peptides, and other molecules.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color Milky White
pH 6 to 7
For Use With (Application) Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats
Content And Storage 4°C
Source Papaya Latex
Quantity 25 mg
Product Type Papain
Molecular Weight (g/mol) 23000
Inhibitors Sulfhydryl groups including heavy Metals, Carbonyl reagents, Aldehydes, Benzoylamidoacetonitrile, AEBSF, antipain, cystatin, E-64, leupeptin, PMSF, TLCK and TPCK
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.