Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G, Sodium Salt, MP Biomedicals™

Catalog No. ICN19453780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mU
100 mU
25 mU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19453780 100 mU
MP219453701 1 mU
MP219453725 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19453780 Supplier MP Biomedicals, Inc Supplier No. 0219453780
Add to Cart
Add to Cart

Penicillin G, Sodium Salt Cell culture grade, Powder

  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae.
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
  • Penicillin G also known as benzylpenicillin.
  • It exert a bacteriocidal as well as a bacteriostatic effect on susceptible bacteria by interfering with the final stages of the synthesis of peptidoglycan, a substance in the bacterial cell wall.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00069666
Synonym Benzylpenicillin sodium salt

Specifications

Physical Form Powder
Loss on Drying 0.001
Potency >1500 μ/mg
Sterility Non-sterile
CAS 69-57-8
Color White
Quantity 100 mU
Validated Application Cell Culture
Molecular Formula C16H17N2NaSO4
Beilstein 3834217
Solubility Information Soluble in water(H2O):100mg/mL.
Molecular Weight (g/mol) 356.38
Grade Cell Culture
Show More Show Less
RTECSNumber XH9800000
storageNote1 Store at +4°C, protect from light.
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.