Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepstatin A

Catalog No. ICN19536801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
250 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19536801 1 mg
ICN19536805 5 mg
ICN19536825 25 mg
ICN19536880 100 mg
ICN19536810 10 mg
ICN19536883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN19536801 Supplier MP Biomedicals™ Supplier No. 0219536801
Add to Cart
Add to Cart

Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.

Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta

C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Molecular Weight (g/mol) 685.91
Name Pepstatin A
Quantity 1 mg
Storage Requirements 0°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.