Learn More
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Specifications
| For Use With (Application) | Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation |
| Molecular Weight (g/mol) | 685.91 |
| Name | Pepstatin A |
| Quantity | 25 mg |
| Storage Requirements | 0°C |
| Form | Powder |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.