Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peptone-Y Powder

Catalog No. MP114018532 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114018532 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114018532 Supplier MP Biomedicals™ Supplier No. 114018532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for yeast media formulations

Yeast cell culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Product Type Peptone
For Use With (Application) For Yeast cell culture
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.