Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peroxidase From Horseradish

Catalog No. ICN15003650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15003650 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15003650 Supplier MP Biomedicals™ Supplier No. 0215003650

missing translation for 'validMainframeMsg'

Useful for tracing neural connections, amplifying weak signals and increasing detectability of a target molecule such as a protein

A plant glycohemoprotein, Horseradish Peroxidase's (HRP's) enzymatic activity is due to the cyclic reduction and oxidation of the iron atom in the hematin group. The iron-containing hemin group is covalently bound to the glycoprotein apoenzyme. HRP readily combines with hydrogen peroxide and the resultant complex can oxidize a wide variety of chromogenic hydrogen donors. It is composed of seven isozymes, all containing photohemin IX as prosthetic group.

  • Specific activity: ≥60 purpurogallin units/mg
  • Extinction coefficient: EmM = 100 (403 nm)(Lit.)
  • Presentation: brown powder
  • Isoelectric Point: 7.2(Lit.)
  • Optimum pH: 7.0(Lit.)
  • Unit definition: amount of enzyme causing the production of 1mg of purpurogallin in 20 seconds at 20°C

RZ: ≥0.6 RZ Definition: The ratio of the absorbance at 403nm to the absorbance at 275nm. This value is an expression of the ratio of hemin to protein content.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown
pH 7
For Use With (Application) ELISA, immunoblotting, immunohistochemistry
Content And Storage -20°C
Source Horseradish
Quantity 50 kU
Product Type Peroxidase
Molecular Weight (g/mol) 44000
Inhibitors p-Aminobenzoic Acid, Azide, Cyclopropanone, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite and Vanadate and Divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.