Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peroxidase, From Horseradish

Catalog No. ICN15003810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15003810 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15003810 Supplier MP Biomedicals™ Supplier No. 0215003810

missing translation for 'validMainframeMsg'

From Horseradish

Peroxidase, [9003-99-0]

  • Lactoperoxidase
  • A salt-free, freeze dried powder
  • Activity: greater than 100 purpurogallin units/mg solid.
  • Unit Definition: That amount of enzyme causing the production of one milligram of purpurogallin in 20 seconds at 20°C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 300 U/mg
Color Reddish Brown
pH 6.0 to 6.5
For Use With (Application) ELISA, immunoblotting, immunohistochemistry
Content And Storage 0°C
Source Horseradish
Quantity 10 kU
Product Type Peroxidase
Inhibitors Cyanide, Sulfide, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-Cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite, Vanadate and the Divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.