Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peroxidase from Horseradish Roots

Catalog No. ICN364514 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN364514 1 g
ICN364511 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN364514 Supplier MP Biomedicals™ Supplier No. 08364514
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for production of peroxidase conjugated antisera

  • From Horseradish Roots
  • Activity: ≥200 units/mg

TRUSTED_SUSTAINABILITY

Specifications

Color Brown to Red Brown
For Use With (Application) Production of Peroxidase Conjugated Antisera, ELISA, immunohistochemistry, western blotting
Content And Storage -20°C
Source Horseradish roots
Quantity 1 g
Product Type Peroxidase
Inhibitors Sodium Azide, Cyanide, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfide, Vanadate, p-aminobenzoic Acid and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+ and Pb2+ ions
Form Lyophilized Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.