Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Phosphodiesterase I from Crotalus adamanteus

Catalog No. ICN10097880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10097880 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10097880 Supplier MP Biomedicals™ Supplier No. 0210097880
Add to Cart
Add to Cart

Used to breaks phosphodiester bonds. It is a membrane-bound glycoprotein that is used to catalyze the hydrolysis of various nucleotide polyphosphates.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥20 μ/mg
pH 9.8 to 10.4.5
For Use With (Application) Used to breaks phosphodiester bonds, catalyze the hydrolysis of various Nucleotide polyphosphates
Content And Storage -20°C
Source Crotalus adamanteus venom
Quantity 100 U
Product Type Phosphodiesterase I
Molecular Weight (g/mol) 115000
Inhibitors Gluctathione, Cysteine and Ascorbic Acid
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.