Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Phospholipase C, Phosphatidylinositol-Specific

Catalog No. ICN19568505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19568505 5 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19568505 Supplier MP Biomedicals™ Supplier No. 0219568505

From Bacillus cereus

Purity: >95% by SDS-PAGE. Activity: 600U/mg protein. Unit Definition: One unit is the amount of enzyme that converts 1 μmol of substrate to product per minute at 37°C., pH 7.5. May be used to release phosphatidylinositol (PI)-linked proteins from membranes. Supplied in 50mM TEA, ph 7.5, 10mM EDTA, 10mM NaN3.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥600 U/mg
Color Clear
Purity >95% by SDS-Page
For Use With (Application) Release Phosphatidylinositol (PI) - linked Proteins from membranes, cleaves the Phospholipid Phosphatidylinositol (PI) into two molecules
Storage Buffer ≈45U/mL in 50mM TEA pH 7.5, 10mM EDTA, 10mM Sodium Azide
Content And Storage -20°C
Source Bacillus cereus
Quantity 5 U
Product Type Phospholipase C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.