Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protamine Sulfate

Catalog No. ICN19472925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10,000 mg
100,000 mg
1000 mg
25000 mg
5000 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472925 25000 mg
ICN19472901 1000 mg
ICN19472905 5000 mg
ICN19472910 10,000 mg
ICN19472980 100,000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19472925 Supplier MP Biomedicals™ Supplier No. 0219472925
Add to Cart
Add to Cart

From salmon sperm

Cell Culture Reagent. Highly purified. Nitrogen: Approx. 22.5%. Hygroscopic white powder.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Powder
For Use With (Application) Antiheparin activity, refining of vaccines, removal of DNA from Protein samples
Product Type Protamine Sulfate from Salmon Sperm
Quantity 25000 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.