Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protein A, Fc Binding Grade

Catalog No. ICN15389105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
20 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15389105 5 mg
ICN15389101 1 mg
ICN15389120 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15389105 Supplier MP Biomedicals™ Supplier No. 0215389105
Add to Cart
Add to Cart

Fc Binding Grade, White Lyophilized Powder. Routinely used for immunoprecipitation assays.

  • Binding capacity 12-15mg of human lgG/mg solid
  • Carboxy truncated material containing 301 amino acid residues
  • White lyophilized powder
  • Source: Staphlococcus aureus , Cowan strain-1

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 12 to 15 mg of Human lgG/mg solid
For Use With (Application) For detection of immunglobulins, purification
Content And Storage -20°C
Quantity 5 mg
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.