Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protexidase Peroxidase Protective Buffer

Catalog No. ICN980631 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN980631 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN980631 Supplier MP Biomedicals™ Supplier No. 08980631
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stabilizes high dilutions of HRP-antibody conjugates

  • Peroxidase Protective Buffer which stabilizes HRP conjugates.
  • This protective buffer is compatible with common blocking agents such as BSA, skim milk (Blotto), Tween-20 and normal serum.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Content And Storage 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.