Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Prothrombin (Factor II) from Human Plasma

Catalog No. ICN19491402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19491402 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19491402 Supplier MP Biomedicals™ Supplier No. 0219491402
Add to Cart
Add to Cart

Percursor to α-thrombin

Supplied in 50% glycerol/H2O.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5v/v
Formulation Supplied in 50% Glycerol/Water
Molecular Weight (g/mol) 72000
Name Prothrombin Human
Quantity 2 mg
Source Human Plasma
Storage Requirements -20°C
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.