Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pseudomonas Agar F

Catalog No. ICN1008817 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1008817 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1008817 Supplier MP Biomedicals™ Supplier No. 091008817
Add to Cart
Add to Cart

Used with glycerol (cat. no. 1004417) for the isolation, cultivation and differentiation of Pseudomonas aeruginosa on the basis of phycocyanin (pigment) production

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Form Dehydrated
Media Type Pseudomonas Agar F
Product Type Agar
Formulation Polipeptone 20g/L, Magnesium Sulfate 1.5g/L, Dipotassium Phosphate 1.5g/L, Agar 14g/L
For Use With (Application) Isolation, cultivation and differentiation of Pseudomonas aeruginosa
Quantity 500 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.