Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pyruvate Kinase from Rabbit Muscle

Catalog No. ICN15199905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100,000 U
25,000 U
50,000 U
5000 U
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15199905 5000 U
ICN15199925 25,000 U
ICN15199950 50,000 U
ICN15199980 100,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15199905 Supplier MP Biomedicals™ Supplier No. 0215199905
Add to Cart
Add to Cart

≥200u/mg protein, White Crystalline Powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥200 U/mg
Color Clear
pH 7.5
For Use With (Application) Used as a coupling Enzyme in conjunction with Lactic Dehydrogenase in quantifying ADP and the activity of Enzymes that catalyze the formation of ADP
Content And Storage -20°C
Source Rabbit muscle
Quantity 5000 U
Product Type Pyruvate Kinase
Molecular Weight (g/mol) 237000
Inhibitors Activity is inhibited by Ca2+, Fluorophosphate inhibits the Enzyme reaction competitively with respect to PEP , ATP inhibits the reaction by removal of Mg2+ from the Substrate MgADP-
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.