Learn More
MP Biomedicals™ Quick-Clean SPIN Filters (Includes Catch Tubes)
Shop All MP Biomedicals, Inc ProductsDescription
- Spin Filters with catch tubes for 50 preps
- Strore at room temperature
Specifications
Specifications
| Content And Storage | Store at 2-8°C |
| Quantity | 50 Preps |
| Type | Spin Filter |
Safety and Handling
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.