Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RNA Matrix

Catalog No. MP111007204 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111007204 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP111007204 Supplier MP Biomedicals™ Supplier No. 111007204

For use in Nucleic Acid Isolation and Purification

  • White Opaque Solution
  • For Research Use Only (RUO)
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.