Learn More
Description
- Kit is designed for rapid isolation and purification of double-stranded plasmid DNA from bacterial cultures. Theprocess yields pure plasmid DNA that is suitable for enzymatic manipulations including DNA sequencing, restriction enzymedigestion, in vitro transcription and PCR.
- For research use only
Specifications
Specifications
| For Use With (Application) | For rapid isolation, purification of double-stranded plasmid DNA from bacterial culture |
| Includes | Pre-Lysis, Alkaline Lysis, Neutralizing, Glassmilk spin buffers, wash soln. concentrate |
| No. of Reactions | 120 Preps |
| Product Type | RPM Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.