Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SC-HIS-LEU-TRP-URA, Powder

Catalog No. MP114440022 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114440022 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114440022 Supplier MP Biomedicals™ Supplier No. 114440022
Add to Cart
Add to Cart

Synthetic Complete Supplement Mixture (SC) of amino acids for S. cerevisiae growth media

Formulation

  • Add 1.57g of SC-His-Leu-Trp-Ura/L of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation DOB or DOBA of 1.57g/L
For Use With (Application) Yeast cell culture, gene expression, transcription regulation
Quantity 30 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.