Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SC-LEU, Powder

Catalog No. MP114410322 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
30g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114410322 30g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114410322 Supplier MP Biomedicals™ Supplier No. 114410322
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Complete Supplement Mixture (SC) of amino acids for S. cerevisiae growth media

Formulation

  • Add 1.8g of SC-Leu to each liter of DOB or DOBA
  • Autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 21mg/L, L-Alanine 86.5mg/L, L-Arginine HCl 85.6mg/L, L-Asparagine 85.6mg/L, L-Aspartic Acid 85.6mg/L, L-Cysteine HCl 85.6mg/L, Glutamine 85.6mg/L, L-Glutamic Acid 85.6mg/L, Glycine 85.6mg/L, L-Histidine HCl 85.6mg/L, Myo-Inositol 85.6mg/L, L-Isoleucine 85.6mg/L, L-Lysine HCl 85.6mg/L, L-Methionine 85.6mg/L, Para-Aminobenzoic Acid 8.6mg/L, L-Phenylalanine 85.6mg/L, L-Proline 85.6mg/L, L-Serine 85.6mg/L, L-Threonine 85.6mg/L, L-Tryptophan 85.6mg/L, L-Tyrosine 85.6mg/L, Uracil 85.6mg/L, L-Valine 85.6mg/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 30 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.