Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SC-LEU-TRP, Powder

Catalog No. MP114420622 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114420622 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114420622 Supplier MP Biomedicals™ Supplier No. 114420622
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Double dropout formulation of Synthetic Complete Supplement Mixture (SC) of amino acids for S. cereviseae growth media

Formulation

  • Add 1.7g of SC to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisia

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation DOB or DOBA of 1.7g/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 30 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.