Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SD MEDIUM-HIS-LEU-TRP, Powder

Catalog No. MP114830065 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114830065 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114830065 Supplier MP Biomedicals™ Supplier No. 114830065
Add to Cart
Add to Cart

Synthetic defined yeast media with all necessary components for selective growth of Saccharomyces cerevisiae

  • Each SD formulation contains the appropriate CSM-based amino acid mixture and 1.7g/liter YNB, 5 g/liter Ammonium Sulfate, and 20g/liter Dextrose
  • Triple dropout formulation
  • Pouches

Formulation

  • Add contents of the pouch (13.7g) to 500mL of distilled, deionized water
  • Add agar 17g/L if pouring plates
  • Autoclave at 121°C for 15 minutes for complete synthetic defined drop-out medium for the growth of S.cerevisiae

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L, 0.62g/L CSM-HIS-LEU-TRP
For Use With (Application) For Yeast cell culture, gene expression, transcription regulation, growth of Saccharomyces cerevisiae, selection of Yeast transformants, confirmation of auxotrophic phenotypes
Packaging Type Pouch
Quantity 10 x 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.