Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SD Medium-LEU, Powder

Catalog No. MP114811075 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 500 mL
10 x 1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114811075 10 x 1000 mL
MP114811065 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP114811075 Supplier MP Biomedicals™ Supplier No. 114811075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Single dropout formulation of Synthetic Defined (SD) Yeast Media which includes all necessary components for selective growth of S. cereviseae

  • Each SD formulation contains the appropriate CSM-based amino acid mixture and 1.7g/liter YNB, 5g/liter Ammonium Sulfate, and 20g/liter Dextrose

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L, CSM-Leu 0.69g/L
For Use With (Application) For confirming auxotrophic phenotypes and selecting Yeast transformants
Includes All necessary components for selective growth of S. cereviseae
Packaging Type Pouch
Quantity 10 x 1000 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.