Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Streptokinase

Catalog No. p-4605383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
50 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10111410 10 kU
ICN10111450 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10111410 Supplier MP Biomedicals™ Supplier No. 0210111410
Add to Cart
Add to Cart

Catalyzes hydrolysis of amide linkages

Forms a 1:1 complex with plasminogen which is then converted to streptokinase-plasmin. Both streptokinase-plasminogen and streptokinase-plasmin complexes can activate plasminogen.

  • Can be used as a thrombolytic agent in therapy of ischemic stroke
  • From: Lancefield group C strain of Streptococcus
  • Presentation: White lyophilized powder
  • Unit definition: Amount of enzyme that will liquefy a standard clot of fibrinogen, plasminogen and thrombin in 10 minutes at 37°C and pH 7.5
  • Specific activity: >5,000 units/mg dry weight
  • Solubility: Soluble in water

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥5000 U/mg
Color White
For Use With (Application) Used as a thrombolytic agent in the therapy of ischemic stroke
Content And Storage -20°C
Source Streptococcus species, lancefield group C strain
Quantity 10 kU
Product Type Streptokinase
Molecular Weight (g/mol) 47400
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.