Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Superoxide Dismutase

Catalog No. ICN19011775 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 kU
3 kU
30 kU
75 kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19011775 75 kU
ICN19011703 3 kU
ICN19011730 30 kU
ICN19011715 15 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19011775 Supplier MP Biomedicals™ Supplier No. 0219011775
Add to Cart
Add to Cart

From bovine erthrocytes

Activity: 3500U/mg. Unit Definition: That amount of enzyme causing a 50% inhibition in the rate of assay based on the method of McCord, J.M. and Fridovich, J. Biol. Chem., 1: 6049, (1969). Lyophilized salt-free powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥0.900 mg protein/mg solid
Color Pale Blue
For Use With (Application) Superoxide Dismutase plays a critical role in the defense of cells against the toxic effects of Oxygen radicals
Content And Storage -20°C
Source Bovine erythrocytes
Quantity 75 kU
Product Type Superoxide Dismutase
Molecular Weight (g/mol) ≈32500
Form Lyophilized Powder
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.