Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Terrific Broth, Dehydrated Media

Catalog No. MP113045032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113045032 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113045032 Supplier MP Biomedicals, Inc Supplier No. 113045032
Add to Cart
Add to Cart

For the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Grade Molecular biology
Product Type Microbiology Media
Formulation Powder, Part 1: Yeast extract 24g/L, Tryptone 12g/L, liquid, Part 2: Potassium phosphate Salts and Glycerol in sterile solution
For Use With (Application) For growth of Escherichia coli
Includes Part 1 powder and Part 2 liquid
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.