Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Terrific Broth II, Dehydrated Media in Pouches
GSA_VA

Catalog No. MP113046075 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113046075 10 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113046075 Supplier MP Biomedicals™ Supplier No. 113046075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich medium for growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Dehydrated
Grade Molecular Biology
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Sodium Chloride and Vitamins
For Use With (Application) For growth of Escherichia coli
Includes 10 × 50g pouches
Packaging Type Pouch
Quantity 10 x 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.