Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Terrific Broth

Catalog No. ICN1010417 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1010417 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1010417 Supplier MP Biomedicals™ Supplier No. 091010417
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the propagation of recombinant E. coli strains.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Media Type Terrific Broth
Product Type Microbiology Media
Formulation Tryptone 12g/L, Yeast extract 24g/L, Monopotassium Phosphate 2.2g/L, Dipotassium Phosphate 9.4g/L
For Use With (Application) Cultivation of molecular genetic strains of Escherichia coli and increased plasmid yield
Quantity 500 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.