Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Supplied in 50% glycerol/H2O.
Specifications
Specifications
| Color | Clear |
| For Use With (Application) | Used for site specific cleavage of recombinant fusion Proteins containing an accessible Thrombin recognition site for removal of affinity tags |
| Content And Storage | -20°C |
| Source | Bovine Plasma |
| Quantity | 1 mg |
| Product Type | alpha-Thrombin |
| Molecular Weight (g/mol) | 36700 |
| Form | Solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?