Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ α-Thrombin from Bovine Plasma

Catalog No. p-3563283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19492191 1 mg
ICN19492182 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19492191 Supplier MP Biomedicals™ Supplier No. 0219492191
Add to Cart
Add to Cart

Highly specific serine protease

Supplied in 50% glycerol/H2O.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Application) Used for site specific cleavage of recombinant fusion Proteins containing an accessible Thrombin recognition site for removal of affinity tags
Content And Storage -20°C
Source Bovine Plasma
Quantity 1 mg
Product Type alpha-Thrombin
Molecular Weight (g/mol) 36700
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.