Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Thrombin from Bovine Plasma

Catalog No. ICN15416380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416380 100 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15416380 Supplier MP Biomedicals™ Supplier No. 0215416380
Add to Cart
Add to Cart

Used for cell biology and analysis

Prepared from fresh bovine plasma by an ion exchange method. Fibrinogen is removed from the prothrombin complex by precipitation. The concentrated prothrombin complex is then activated by beef thromboplastin and calcium.

  • Format: White powder
  • High purity grade
  • Molecular Weight: ∽33,500d
  • Specific Activity: ≥1500 NIH Units/mg protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈10%
Color White
pH 7 ±0.5
For Use With (Application) Research tool, medicine, surgery, food industry
Content And Storage -20°C
Source Bovine
Quantity 100 kU
Product Type Thrombin
Molecular Weight (g/mol) ≈32500
Inhibitors AEBSF, PMSF, Antithrombin, TLCK, 3,4-dichlorocoumarin, Leupeptin
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.