Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas

Catalog No. ICN10313983 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10313983 250 g
ICN10313925 25 g
ICN10313980 100 g
ICN10313990 500 g
ICN10313991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10313983 Supplier MP Biomedicals™ Supplier No. 0210313983
Add to Cart
Add to Cart

From Hog Pancreas

Trypsin, 1 - 250, [9002-07-7]

  • Parenzymol
  • Dry powder: stable
  • Protease act: min 250,000 USP units/gm

Dry Powder

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 24000
Name Trypsin
pH Range ∽8
Quantity 250 g
Source Hog
Storage Requirements 4°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.