Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trypsin Inhibitor, From Chicken Eggwhites

Catalog No. ICN10061201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
0.5 g
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10061201 1 g
ICN1006122 250 mg
ICN1006125 0.5 g
ICN10061205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10061201 Supplier MP Biomedicals™ Supplier No. 0210061201
Add to Cart
Add to Cart

Mucoprotein and antitryptic factor of egg white described by Lineweaver and Murray1. Supplied as a dried powder. A glycoprotein.

Trypsin Inhibitor, [9035-81-8]

  • Essentially Salt-free
  • Lyophilized
  • 1 mg inhibits 1.0-3.0 mg of trypsin
  • CAS # : 9035-81-8
  • Activity: 1mg inhibits approxiately 2.5 to 3.0mg of trypsin.

1. Lineweaver and Murray, J. Biol. Chem., v. 171, 565 (1947)

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 28500
Name Trypsin Inhibitor
Quantity 1 g
Source Chicken Egg White
Storage Requirements 4°C
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.