Learn More
Description
Description:
MP Wheat Germ is raw and has not been heat treated in any manner. Contains wheat germ, bran and middlings.
Physical Description:
Brown flakes/powder
Specifications
Specifications
| Product Type | Animal Diet |
| Format | Flakes |
| For Use With (Application) | In molecular biology, wheat germ extract is used to carry out in-vitro translation experiments, as the plant embryo contains all the macromolecular components necessary for translating mRNA into amino Acids, but contains relatively low levels of its own mRNA |
| Quantity | 11.33 kg |
| Storage Requirements | 15°C to 30°C |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.