Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Wheat Germ Agglutinin

Catalog No. ICN15226680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15226680 100 mg
ICN15226625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15226680 Supplier MP Biomedicals™ Supplier No. 0215226680/DEL

MP Biomedicals™ Wheat Germ Agglutinin has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers.

  • Less than 4 mg/mL will agglutinate human type O erythrocytes, and less than 1 mg/mL will agglutinate neuraminidase-treated erythrocytes
  • Contains no protein-bound carbohydrate
TRUSTED_SUSTAINABILITY

Specifications

Color White to Pale Yellow
Quantity 100 mg
Source Triticum vulgaris
Content And Storage -20°C
Form Lyophilized
For Use With (Application) Proteomics, immune system regulation
Product Type Lectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.